Nickel(II) acetate tetrahydrate, 98+%, Thermo Scientific Chemicals
Nickel(II) acetate tetrahydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel(II) acetate tetrahydrate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
100 g
500 g
Catalog number A13026.0E
also known as A13026-0E
Price (USD)/ Each
241.65
Online exclusive
268.00 
Save 26.35 (10%)
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
241.65
Online exclusive
268.00 
Save 26.35 (10%)
Add to cart
Nickel(II) acetate tetrahydrate, 98+%, Thermo Scientific Chemicals
Catalog numberA13026.0E
Price (USD)/ Each
241.65
Online exclusive
268.00 
Save 26.35 (10%)
-
Add to cart
Chemical Identifiers
CAS6018-89-9
IUPAC Namenickel(2+) diacetate tetrahydrate
Molecular FormulaC4H14NiO8
InChI KeyOINIXPNQKAZCRL-UHFFFAOYSA-L
SMILESO.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (unspecified)≥98.0 to ≤102.0% (based on elemental Ni)
Complexometric TitrationNickel Ni: 23.1-24.05%
Appearance (Color)Green
Nickel(II) acetate tetrahydrate is used in electroplating. It is also used in the preparation of polynuclear-nickel polyoxotungstate cluster compounds, which are used in designing molecular magnets. Further, it is used as a catalyst and mordant for dyeing textiles, as a sealant for anodized aluminum and to make other nickel compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nickel(II) acetate tetrahydrate is used in electroplating. It is also used in the preparation of polynuclear-nickel polyoxotungstate cluster compounds, which are used in designing molecular magnets. Further, it is used as a catalyst and mordant for dyeing textiles, as a sealant for anodized aluminum and to make other nickel compounds.

Solubility
Soluble in water and methanol. Insoluble in diethyl ether and n-octanol.

Notes
Hygroscopic. Incompatible with oxidizing agents, strong acids and strong bases.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. In combination with NaH and the Na derivative of 3-Hydroxy-1-methyl piperidine, L10829 , can be used for desufurization of thioethers, showing dramatic rate enhancement compared with other systems: Tetrahedron Lett., 39, 8987 (1998).
  2. Das, M.; Harms, K.; Ghosh, B. N.; Rissanen, K.; Chattopadhyay, S. Bis(μ-tetrazolato-NN') bridged dinuclear nickel(II) Schiff base complexes: Tandem synthesis, structure and self assembly. Polyhedron 2015, 87, 286-292.
  3. Sivakumar, P.; Nayak, P. K.; Markovsky, B.; Aurbach, D.; Gedanken, A. Sonochemical synthesis of LiNi0.5Mn1.5O4 and its electrochemical performance as a cathode material for 5V Li-ion batteries. Ultrason. Sonochem. 2015, 26, 332-339.