Thermo Scientific Chemicals

3,4-Dimethylphenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A13027.09
10 g, Each
Thermo Scientific Chemicals

3,4-Dimethylphenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A13027.09
10 g, Each
Quantity
Catalog number: A13027.09
also known as A13027-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
60481-51-8
IUPAC Name
hydrogen (3,4-dimethylphenyl)hydrazine chloride
Molecular Formula
C8H13ClN2
InChI Key
YYMIOVAEQIEPET-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CC1=CC=C(NN)C=C1C
Appearance (Color)
Pale cream to cream to brown
Form
Powder or flakes
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%

Description

3,4-Dimethylphenylhydrazine hydrochloride, is used as a reactant with ethyl 3-ethoxy-2-ethoxycarbonylprop-2-enoate to produce 2-(3,4-dimethyl-phenyl)-3-oxo-2,3-dihydro-1H-pyrazole-4-carboxylic acid ethyl ester. This reaction will need reagent K2CO3, and the menstruum ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethylphenylhydrazine hydrochloride, is used as a reactant with ethyl 3-ethoxy-2-ethoxycarbonylprop-2-enoate to produce 2-(3,4-dimethyl-phenyl)-3-oxo-2,3-dihydro-1H-pyrazole-4-carboxylic acid ethyl ester. This reaction will need reagent K2CO3, and the menstruum ethanol.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text