Thermo Scientific Chemicals

Methyl methacrylate, 99%, stab., Thermo Scientific Chemicals

Catalog number: A13030.AE
100 mL, Each
Thermo Scientific Chemicals

Methyl methacrylate, 99%, stab., Thermo Scientific Chemicals

Catalog number: A13030.AE
100 mL, Each
Quantity
Catalog number: A13030.AE
also known as A13030-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
80-62-6
IUPAC Name
methyl 2-methylprop-2-enoate
Molecular Formula
C5H8O2
InChI Key
VVQNEPGJFQJSBK-UHFFFAOYSA-N
SMILES
COC(=O)C(C)=C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%
Comment
Acidity: ≤0.005% (as methacrylic acid)
Identification (FTIR)
Conforms

Description

Methyl methacrylate is used as a monomer in the manufacturing of poly(methyl methacrylate) (PMMA). It is also used in the manufacturing of the co-polymer methyl methacrylate-butadiene-styrene (MBS), which is a modifier for polyvinyl chloride. It plays an important role as the cement used in total hip replacements as well as total knee replacements. It is also involved in the preparation of other methacrylates viz. ethyl methacrylate (EMA), butyl methacrylate (BMA) and 2-ethyl hexyl methacrylate (2-EHMA) and methacrylic acid (MAA), which finds application as an intermediate in the manufacture of coating polymers and textile applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl methacrylate is used as a monomer in the manufacturing of poly(methyl methacrylate) (PMMA). It is also used in the manufacturing of the co-polymer methyl methacrylate-butadiene-styrene (MBS), which is a modifier for polyvinyl chloride. It plays an important role as the cement used in total hip replacements as well as total knee replacements. It is also involved in the preparation of other methacrylates viz. ethyl methacrylate (EMA), butyl methacrylate (BMA) and 2-ethyl hexyl methacrylate (2-EHMA) and methacrylic acid (MAA), which finds application as an intermediate in the manufacture of coating polymers and textile applications.

Solubility
Slightly miscible with water.

Notes
Incompatible with oxidizing agents, peroxides, amines, bases, acids, reducing agents and halogens.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text