Thermo Scientific Chemicals

Phloroglucinol dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: A13034.36
500 g, Each
Thermo Scientific Chemicals

Phloroglucinol dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: A13034.36
500 g, Each
Quantity
Catalog number: A13034.36
also known as A13034-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6099-90-7
IUPAC Name
benzene-1,3,5-triol dihydrate
Molecular Formula
C6H10O5
InChI Key
MPYXTIHPALVENR-UHFFFAOYSA-N
SMILES
O.O.OC1=CC(O)=CC(O)=C1
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥97.5%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
18-24%
Appearance (Color)
White to cream to brown

Description

Phloroglucinol dihydrate is utilized in the preparation of active pharmaceutical ingredients such as flopropione. It acts as a reagent for the Tollens’ test for pentoses. It is mixed with hydrochloric acid and plays an important role for the detection of lignin (Wiesner test). It is involved in the analysis and qualitative detection of free hydrochloric acid in gastric juice by using Gunzburg reagent. Further, it serves as a coupling agent in printing. It is also employed as a reagent for pentoses, pentosans, aldehydes and as a decalcifier of bone specimens in microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phloroglucinol dihydrate is utilized in the preparation of active pharmaceutical ingredients such as flopropione. It acts as a reagent for the Tollens’ test for pentoses. It is mixed with hydrochloric acid and plays an important role for the detection of lignin (Wiesner test). It is involved in the analysis and qualitative detection of free hydrochloric acid in gastric juice by using Gunzburg reagent. Further, it serves as a coupling agent in printing. It is also employed as a reagent for pentoses, pentosans, aldehydes and as a decalcifier of bone specimens in microscopy.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with acid chlorides, acid anhydrides, bases and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text