3-Aminopropionitrile is used for organic synthesis and as pharmaceutical intermediates. As an intermediate used in the manufacture of beta-alanine and pantothenic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Aminopropionitrile is used for organic synthesis and as pharmaceutical intermediates. As an intermediate used in the manufacture of beta-alanine and pantothenic acid.
Solubility
Soluble in water.
Notes
It is air and moisture sensitive. Incompatible with oxidizing agents, acids and heat.
RUO – Research Use Only
Ole-Anders Braathen; K.M.Marstokk; Harald Mollendal. Microwave spectrum, conformational equilibrium, intramolecular hydrogen bonding and centrifugal distortion of 3-aminopropionitrile. Acta Chem. Scand. A 1983, 37, 493-501
Edwin M.Smolin; L.Clair Beegle. Continuous high pressure synthesis of 3-aminopropionitrile. Ind. Eng. Chem. 1958, 50, (8),1115-1118