General Description
• N-Acetyl-D-glucosamine is the acetylated form of glucosamine, a glucose derivative
• It has a role as a prokaryotic metabolism and monomer used in chitin, hyaluronic acid, and polymers within the cell wall
Application
• N-Acetyl-D-Glucosamine can be used for molecular binding studies; research has shown that it attaches to certain microbes and their pathogens
• It is useful for biochemical characterization studies of related glucosamines
General References: