Thermo Scientific Chemicals

2,4,6-Trimethylaniline, 98%, Thermo Scientific Chemicals

Catalog number: A13049.0E
2500 g, Each
Thermo Scientific Chemicals

2,4,6-Trimethylaniline, 98%, Thermo Scientific Chemicals

Catalog number: A13049.0E
2500 g, Each
Quantity
Catalog number: A13049.0E
also known as A13049-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
88-05-1
IUPAC Name
2,4,6-trimethylanilinium chloride
Molecular Formula
C9H14ClN
InChI Key
WUYJXWRFOUCHEB-UHFFFAOYSA-N
SMILES
[Cl-].CC1=CC(C)=C([NH3+])C(C)=C1
Assay (GC)
≥97.5%
Appearance (Color)
Clear, pale yellow to orange or brown
Refractive Index
1.5490-1.5530 @20?C
Identification (FTIR)
Conforms
Form
Liquid

Description

2,4,6-Trimethylaniline is useful for the preparation of Grubbs' catalyst. It is used as a precursor to dyes. It undergoes condensation reaction with glyoxal to prepare glyoxal-bis(mesitylimine). It is also involved in the preparation of 1,3-diketimines ligands by condensation with 1,3-diketones. Further, it is used for the production of Acid Blue 129 dye, which is useful in histochemistry studies. In addition to this, it serves as a building block to various bulky ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethylaniline is useful for the preparation of Grubbs′ catalyst. It is used as a precursor to dyes. It undergoes condensation reaction with glyoxal to prepare glyoxal-bis(mesitylimine). It is also involved in the preparation of 1,3-diketimines ligands by condensation with 1,3-diketones. Further, it is used for the production of Acid Blue 129 dye, which is useful in histochemistry studies. In addition to this, it serves as a building block to various bulky ligands.

Solubility
Slightly miscible with carbon tetrachloride. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text