5-Decyne, 98%, Thermo Scientific Chemicals
5-Decyne, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Decyne, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number A13050.18
also known as A13050-18
Price (USD)/ Each
221.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
221.00
Add to cart
5-Decyne, 98%, Thermo Scientific Chemicals
Catalog numberA13050.18
Price (USD)/ Each
221.00
-
Add to cart
Chemical Identifiers
CAS1942-46-7
IUPAC Namedec-5-yne
Molecular FormulaC10H18
InChI KeyJWBQJUFCNOLNNC-UHFFFAOYSA-N
SMILESCCCCC#CCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.4315-1.4355 @ 20?C
FormLiquid
5-Decyne is used as APIs and intermediates. It is also used for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Decyne is used as APIs and intermediates. It is also used for chemical research.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Toshio Masuda; Yoshinori Kuwane; Toshinobu Higashimura. Polymerization of symmetrical dialkylacetylenes (3-Hexyne, 4-Octyne, and 5-Decyne) by WCl6·Ph4Sn and MoCl5·Ph4Sn. Polymer Journal. 1981, 13, (3),301-303