Thermo Scientific Chemicals

4-Hydroxy-3,5-dimethylbenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A13051.06
5 g, Each
Thermo Scientific Chemicals

4-Hydroxy-3,5-dimethylbenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A13051.06
5 g, Each
Quantity
Catalog number: A13051.06
also known as A13051-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4919-37-3
IUPAC Name
4-hydroxy-3,5-dimethylbenzoate
Molecular Formula
C9H9O3
InChI Key
OMNHTTWQSSUZHO-UHFFFAOYSA-M
SMILES
CC1=CC(=CC(C)=C1O)C([O-])=O
Appearance (Color)
White to cream to pink or pale brown
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Melting Point (clear melt)
221.0-227.0?C

Description

4-hydroxy-3,5-dimethylbenzoic acid is widely used as a reagent for potent TTR amyloidogenesis inhibitors. It plays an important role as an intermediate in agrochemicals and dyestuffs. It is also used in the preparation of mexiletine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-hydroxy-3,5-dimethylbenzoic acid is widely used as a reagent for potent TTR amyloidogenesis inhibitors. It plays an important role as an intermediate in agrochemicals and dyestuffs. It is also used in the preparation of mexiletine derivatives.

Solubility
Soluble in methanol.

Notes
Keep away from heat and avoid excessive light. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text