Thermo Scientific Chemicals

2-Aminopyrazine, 99%, Thermo Scientific Chemicals

Catalog number: A13052.06
5 g, Each
Thermo Scientific Chemicals

2-Aminopyrazine, 99%, Thermo Scientific Chemicals

Catalog number: A13052.06
5 g, Each
Quantity
Catalog number: A13052.06
also known as A13052-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5049-61-6
IUPAC Name
pyrazin-2-amine
Molecular Formula
C4H5N3
InChI Key
XFTQRUTUGRCSGO-UHFFFAOYSA-N
SMILES
NC1=CN=CC=N1
Appearance (Color)
White to cream to brown
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.5%
Melting Point (clear melt)
116.0-123.0?C

Description

2-Aminopyrazine acts as a raw material in a four-component synthesis of imidazolidines. It is also used as an intermediate in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminopyrazine acts as a raw material in a four-component synthesis of imidazolidines. It is also used as an intermediate in synthetic chemistry.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text