Thermo Scientific Chemicals

Ethyl 3-phenylpropionate, 98+%, Thermo Scientific Chemicals

Catalog number: A13055.0B
1000 g, Each
Thermo Scientific Chemicals

Ethyl 3-phenylpropionate, 98+%, Thermo Scientific Chemicals

Catalog number: A13055.0B
1000 g, Each
Quantity
Catalog number: A13055.0B
also known as A13055-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2021-28-5
IUPAC Name
ethyl 3-phenylpropanoate
Molecular Formula
C11H14O2
InChI Key
JAGZUIGGHGTFHO-UHFFFAOYSA-N
SMILES
CCOC(=O)CCC1=CC=CC=C1
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
>98.0%
Refractive Index
1.4920-1.4960 @ 20?C

Description

Ethyl 3-phenylpropionate is used as an active pharmaceutical ingredient intermediate. It is also used in the synthesis of pyrimidine based inhibitors of cyclin-dependant kinases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-phenylpropionate is used as an active pharmaceutical ingredient intermediate. It is also used in the synthesis of pyrimidine based inhibitors of cyclin-dependant kinases.

Solubility
Miscible with oils and ethanol. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text