Thermo Scientific Chemicals

Ethyl benzoate, 99%, Thermo Scientific Chemicals

Catalog number: A13057.22
100 g, Each
Thermo Scientific Chemicals

Ethyl benzoate, 99%, Thermo Scientific Chemicals

Catalog number: A13057.22
100 g, Each
Quantity
Catalog number: A13057.22
also known as A13057-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
93-89-0
IUPAC Name
ethyl benzoate
Molecular Formula
C9H10O2
InChI Key
MTZQAGJQAFMTAQ-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC=CC=C1
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.5035-1.5065 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid

Description

Ethyl benzoate is used as a perfume scent. It acts as a food flavoring agent. It is an active component of artificial fruit flavors. Further, it is used in cosmetics and personal care products as fragrance ingredients and preservatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl benzoate is used as a perfume scent. It acts as a food flavoring agent. It is an active component of artificial fruit flavors. Further, it is used in cosmetics and personal care products as fragrance ingredients and preservatives.

Solubility
Miscible with methanol and diethyl ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text