Thermo Scientific Chemicals

2-Iodoaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A13059.14
25 g, Each
Thermo Scientific Chemicals

2-Iodoaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A13059.14
25 g, Each
Quantity
Catalog number: A13059.14
also known as A13059-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
615-43-0
Form
Crystals or powder or crystalline powder or needles
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Appearance (Color)
White to cream to orange to brown

Description

2-Iodoaniline is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodoaniline is used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
It is sensitive to light. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text