2-Iodoaniline is used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Iodoaniline is used as an intermediate in organic synthesis.
Solubility
Insoluble in water.
Notes
It is sensitive to light. Incompatible with oxidizing agents.
RUO – Research Use Only
Gozen Bereket; Evrim Hur; Yucel Sahin. Electrodeposition of polyaniline, poly(2-iodoaniline), and poly(aniline-co-2-iodoaniline) on steel surfaces and corrosion protection of steel. Applied Surface Science. 2005, 252, (5),1233-1244
Qiuping Ding; Banpeng Cao; Xianjin Liu; Zhenzhen Zongand; Yi-Yuan Peng. Synthesis of 2-aminobenzothiazole via FeCl3-catalyzed tandem reaction of 2-iodoaniline with isothiocyanate in water. Green Chemistry. 2010, (9),1607-1610
Quinolines may be synthesized by Pd-catalyzed reaction with allyl alcohols: Tetrahedron Lett., 32, 569 (1991).
Reacts with unsymmetrical acetylenes in the presence of Palladium(II) acetate, 10516, to give indoles substituted at the 2- and/or 3-positions: J. Am. Chem. Soc., 113, 6689 (1991):
Under similar conditions, ketones with an ɑ-methylene group also give indole derivatives: J. Org. Chem., 62, 2676 (1997).