Thermo Scientific Chemicals

2-Acetylthiophene, 99%, Thermo Scientific Chemicals

Catalog number: A13062.22
100 g, Each
Thermo Scientific Chemicals

2-Acetylthiophene, 99%, Thermo Scientific Chemicals

Catalog number: A13062.22
100 g, Each
Quantity
Catalog number: A13062.22
also known as A13062-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
88-15-3
IUPAC Name
1-(thiophen-2-yl)ethan-1-one
Molecular Formula
C6H6OS
InChI Key
WYJOVVXUZNRJQY-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=CS1
Assay (GC)
≥98.5%
Appearance (Color)
Clear colorless to yellow to pale brown
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.5640-1.5670 @ 20?C

Description

2-Acetylthiophene is used as an intermediate in the manufacturing of drugs like Tiamonium Iodide, Suprofan, Stepronin, Tenonitrozole, Namirotene, etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylthiophene is used as an intermediate in the manufacturing of drugs like Tiamonium Iodide, Suprofan, Stepronin, Tenonitrozole, Namirotene, etc.

Solubility
Insoluble in water.

Notes
It is sensitive to light. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text