Thermo Scientific Chemicals

Cyclobutanone, 98%, stab. with ca 0.1% BHT, Thermo Scientific Chemicals

Catalog number: A13068.06
5 g, Each
Thermo Scientific Chemicals

Cyclobutanone, 98%, stab. with ca 0.1% BHT, Thermo Scientific Chemicals

Catalog number: A13068.06
5 g, Each
Quantity
Catalog number: A13068.06
also known as A13068-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1191-95-3
Appearance (Color)
Clear or slightly turbid, colorless to pale yellow
Form
Liquid
Assay (GC)
>97.5%
Refractive Index
1.4190-1.4230 @ 20?C

Description

Cyclobutanone is used in preparation of cyclobutane derivatives. It is used as a starting material for aminocyclobutanecarboxylic acid through sulfonyloxiranes. It is also used to prepare dihydro-furan-2-one in presence of polystyrene-bound phenylselenic acid as catalyst. Further, it is involved in the photochemical synthesis of nucleoside analogues such as bicyclic nucleosides and isonucleosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclobutanone is used in preparation of cyclobutane derivatives. It is used as a starting material for aminocyclobutanecarboxylic acid through sulfonyloxiranes. It is also used to prepare dihydro-furan-2-one in presence of polystyrene-bound phenylselenic acid as catalyst. Further, it is involved in the photochemical synthesis of nucleoside analogues such as bicyclic nucleosides and isonucleosides.

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text