Thermo Scientific Chemicals

1,2,4-Trihydroxybenzene, 97%, Thermo Scientific Chemicals

Catalog number: A13069.03
1 g, Each
Thermo Scientific Chemicals

1,2,4-Trihydroxybenzene, 97%, Thermo Scientific Chemicals

Catalog number: A13069.03
1 g, Each
Quantity
Catalog number: A13069.03
also known as A13069-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
533-73-3
IUPAC Name
benzene-1,2,4-triol
Molecular Formula
C6H6O3
InChI Key
GGNQRNBDZQJCCN-UHFFFAOYSA-N
SMILES
OC1=CC=C(O)C(O)=C1
Appearance (Color)
White to grey or pale brown to brown
Form
Powder
Assay (GC)
≥96.0%
Infrared spectrum
Conforms

Description

1,2,4-Trihydroxybenzene is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.It is used as a metabolite of benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,4-Trihydroxybenzene is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.It is used as a metabolite of benzene.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acid chlorides, acid anhydrides, bases, oxidizing agents. Air & Light Sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text