Thermo Scientific Chemicals

2-(1-Piperidinyl)aniline, 98%, Thermo Scientific Chemicals

Catalog number: A13073.06
5 g, Each
Thermo Scientific Chemicals

2-(1-Piperidinyl)aniline, 98%, Thermo Scientific Chemicals

Catalog number: A13073.06
5 g, Each
Quantity
Catalog number: A13073.06
also known as A13073-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39643-31-7
IUPAC Name
2-(piperidin-1-yl)aniline
Molecular Formula
C11H16N2
InChI Key
OYECAJPUPWFCSL-UHFFFAOYSA-N
SMILES
NC1=CC=CC=C1N1CCCCC1
Assay (GC)
≥97.5%
Appearance (Color)
Brown to dark brown
Melting Point (clear melt)
43.0-49.0°C
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text