Thermo Scientific Chemicals

2-(2-Chloroethoxy)ethanol, 99%, Thermo Scientific Chemicals

Catalog number: A13074.18
50 g, Each
Thermo Scientific Chemicals

2-(2-Chloroethoxy)ethanol, 99%, Thermo Scientific Chemicals

Catalog number: A13074.18
50 g, Each
Quantity
Catalog number: A13074.18
also known as A13074-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
628-89-7
IUPAC Name
2-(2-chloroethoxy)ethan-1-ol
Molecular Formula
C4H9ClO2
InChI Key
LECMBPWEOVZHKN-UHFFFAOYSA-N
SMILES
OCCOCCCl
Assay (GC)
≥98.5%
Appearance (Color)
Clear colorless
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4520-1.4550 @ 20?C

Description

2-(2-Chloroethoxy)ethanol is employed in the preparation of o-nitrophenylbromoacetaldehyde bis-2-(2-chloroethoxy)-ethyl acetal, quetiapine and 2-(2-azidoethoxy)ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Chloroethoxy)ethanol is employed in the preparation of o-nitrophenylbromoacetaldehyde bis-2-(2-chloroethoxy)-ethyl acetal, quetiapine and 2-(2-azidoethoxy)ethanol.

Solubility
Miscible with water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text