Thermo Scientific Chemicals

1-Acetylferrocene, 97%, Thermo Scientific Chemicals

Catalog number: A13078.14
25 g, Each
Thermo Scientific Chemicals

1-Acetylferrocene, 97%, Thermo Scientific Chemicals

Catalog number: A13078.14
25 g, Each
Quantity
Catalog number: A13078.14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1271-55-2
IUPAC Name
1-Acetylferrocene
Molecular Formula
C12H12FeO
SMILES
[Fe].c1cccc1.CC(=O)c1cccc1
Molecular Weight (g/mol)
228.07
Appearance (Color)
Orange to orange-brown or red-brown or red
Form
Crystals or crystalline powder and/or lumps
Assay (chromatographic, GC or HPLC)
≥96.0%

Description

1-Acetylferrocene was originally used in military or space field as an additive in rocket propellant, to promote the burning rate. Its ferrocenyl derivative has wide applications to biological and medical fields such as ferrocene-modified beta-lactam because of its physiological activity of anti-malarial, anti-tumor, bactericidal, anti-inflammatory, treatment of anemia, inhibition of enzymatic activity and so on by virtue of its unique structure and diverse properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water. Soluble in most organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text