Resorcinol, 99%, Thermo Scientific Chemicals
Resorcinol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Resorcinol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5000 g
50 g
250 g
1000 g
Catalog number A13080.0I
also known as A13080-0I
Price (USD)/ Each
582.65
Online exclusive
647.00 
Save 64.35 (10%)
-
Add to cart
Quantity:
5000 g
Request bulk or custom format
Price (USD)/ Each
582.65
Online exclusive
647.00 
Save 64.35 (10%)
Add to cart
Resorcinol, 99%, Thermo Scientific Chemicals
Catalog numberA13080.0I
Price (USD)/ Each
582.65
Online exclusive
647.00 
Save 64.35 (10%)
-
Add to cart
Chemical Identifiers
CAS108-46-3
IUPAC Namebenzene-1,3-diol
Molecular FormulaC6H6O2
InChI KeyGHMLBKRAJCXXBS-UHFFFAOYSA-N
SMILESOC1=CC(O)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to pale brown or pink
Identification (FTIR)Conforms (non-U.S. specification)
FormCrystals or powder or crystalline powder or granules or flakes
CommentSpecification differs for U.S. and non-U.S. material where indicated
Assay (GC)≥98.5%
View more
Resorcinol is used in the synthesis of pharmaceuticals and various organic compounds. It is likewise used in plasticizers and diazo dyes. It can be applied in the qualitative determination of ketoses in the Seliwanoff test. Resorcinol-formaldehyde resin has shown useful application in endodontic therapy in russian red. It is an indispensable element for spectrophotometric determination of nitrate concentrations in seawater.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Resorcinol is used in the synthesis of pharmaceuticals and various organic compounds. It is likewise used in plasticizers and diazo dyes. It can be applied in the qualitative determination of ketoses in the Seliwanoff test. Resorcinol-formaldehyde resin has shown useful application in endodontic therapy in russian red. It is an indispensable element for spectrophotometric determination of nitrate concentrations in seawater.

Solubility
Soluble in water, alcohol, ether and glycerol.

Notes
Stable. May discolor on exposure to air or light.
RUO – Research Use Only

General References:

  1. Zhang, J. Z.; Fischer, C. J. A simplified resorcinol method for direct spectrophotometric determination of nitrate in seawater. Marine Chemistry 2006, 99 (1-4), 220-226.
  2. Schwandt, N. W.; Gound, T. G. Resorcinol-Formaldehyde Resin Russian Red Endodontic Therapy. J. Endod. 2003, 29 (7), 435-437.