Thermo Scientific Chemicals

Resorcinol, 99%, Thermo Scientific Chemicals

Catalog number: A13080.30
250 g, Each
Thermo Scientific Chemicals

Resorcinol, 99%, Thermo Scientific Chemicals

Catalog number: A13080.30
250 g, Each
Quantity
Catalog number: A13080.30
also known as A13080-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-46-3
IUPAC Name
benzene-1,3-diol
Molecular Formula
C6H6O2
InChI Key
GHMLBKRAJCXXBS-UHFFFAOYSA-N
SMILES
OC1=CC(O)=CC=C1
Appearance (Color)
White to cream to yellow to pale brown or pink
Identification (FTIR)
Conforms (non-U.S. specification)
Form
Crystals or powder or crystalline powder or granules or flakes
Comment
Specification differs for U.S. and non-U.S. material where indicated
Assay (GC)
≥98.5%

Description

Resorcinol is used in the synthesis of pharmaceuticals and various organic compounds. It is likewise used in plasticizers and diazo dyes. It can be applied in the qualitative determination of ketoses in the Seliwanoff test. Resorcinol-formaldehyde resin has shown useful application in endodontic therapy in russian red. It is an indispensable element for spectrophotometric determination of nitrate concentrations in seawater.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Resorcinol is used in the synthesis of pharmaceuticals and various organic compounds. It is likewise used in plasticizers and diazo dyes. It can be applied in the qualitative determination of ketoses in the Seliwanoff test. Resorcinol-formaldehyde resin has shown useful application in endodontic therapy in russian red. It is an indispensable element for spectrophotometric determination of nitrate concentrations in seawater.

Solubility
Soluble in water, alcohol, ether and glycerol.

Notes
Stable. May discolor on exposure to air or light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text