Thermo Scientific Chemicals

4-Bromo-2-fluorobenzonitrile, 99%, Thermo Scientific Chemicals

Catalog number: A13086.14
25 g, Each
Thermo Scientific Chemicals

4-Bromo-2-fluorobenzonitrile, 99%, Thermo Scientific Chemicals

Catalog number: A13086.14
25 g, Each
Quantity
Catalog number: A13086.14
also known as A13086-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
105942-08-3
IUPAC Name
4-bromo-2-fluorobenzonitrile
Molecular Formula
C7H3BrFN
InChI Key
HGXWRDPQFZKOLZ-UHFFFAOYSA-N
SMILES
FC1=CC(Br)=CC=C1C#N
Solution Test
Clear (5% in Dichloromethane)
Appearance (Color)
White to cream or pale yellow or pale brown
Form
Crystalline powder
Assay (GC)
>98.5%

Description

4-Bromo-2-fluorobenzonitrile is used as a OLED intermediate, Pharmaceutical, electronic and chemical intermediate. It is used in the synthesis of heterocycles and liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-fluorobenzonitrile is used as a OLED intermediate, Pharmaceutical, electronic and chemical intermediate. It is used in the synthesis of heterocycles and liquid crystals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text