Thermo Scientific Chemicals

2-Pyridineacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: A13087.06
5 g, Each
Thermo Scientific Chemicals

2-Pyridineacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: A13087.06
5 g, Each
Quantity
Catalog number: A13087.06
also known as A13087-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2739-97-1
IUPAC Name
2-(pyridin-2-yl)acetonitrile
Molecular Formula
C7H6N2
InChI Key
UKVQBONVSSLJBB-UHFFFAOYSA-N
SMILES
N#CCC1=CC=CC=N1
Appearance (Color)
White to yellow to orange to red to dark brown
Refractive Index
1.5230-1.5300 as melt
Form
Low melting solid (clear liquid as melt)
Assay (GC)
≥96.5%

Description

2-Pyridineacetonitrile is used as a pharmaceutical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Pyridineacetonitrile is used as a pharmaceutical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Miscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text