Thermo Scientific Chemicals

2,6-Di-tert-butyl-p-benzoquinone, 98+%, Thermo Scientific Chemicals

Catalog number: A13091.06
5 g, Each
Thermo Scientific Chemicals

2,6-Di-tert-butyl-p-benzoquinone, 98+%, Thermo Scientific Chemicals

Catalog number: A13091.06
5 g, Each
Quantity
Catalog number: A13091.06
also known as A13091-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
719-22-2
IUPAC Name
2,6-di-tert-butylcyclohexa-2,5-diene-1,4-dione
Molecular Formula
C14H20O2
InChI Key
RDQSIADLBQFVMY-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC(=O)C=C(C1=O)C(C)(C)C
Appearance (Color)
Orange
Assay (GC)
>98.0%
Form
Crystals or crystalline powder
Clarity
5% w/v solution in ethanol, 99.5% will be transparent to almost transparent by JIS K8001 5.2

Description

2,6-Di-tert-butyl-1,4-benzoquinone was used as an antioxidant to study the elimination rate of micro pollutant from storm and waste water. It was also used as a metabolite of butylated hydroxytoluene. It is used as a treatment agent to improve germination and plant health.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Di-tert-butyl-1,4-benzoquinone was used as an antioxidant to study the elimination rate of micro pollutant from storm and waste water. It was also used as a metabolite of butylated hydroxytoluene. It is used as a treatment agent to improve germination and plant health.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text