Thermo Scientific Chemicals

2-Mercapto-1-methylimidazole, 98%, Thermo Scientific Chemicals

Catalog number: A13094.22
100 g, Each
Thermo Scientific Chemicals

2-Mercapto-1-methylimidazole, 98%, Thermo Scientific Chemicals

Catalog number: A13094.22
100 g, Each
Quantity
Catalog number: A13094.22
also known as A13094-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
60-56-0
IUPAC Name
1-methyl-2,3-dihydro-1H-imidazole-2-thione
Molecular Formula
C4H6N2S
InChI Key
PMRYVIKBURPHAH-UHFFFAOYSA-N
SMILES
CN1C=CNC1=S
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Melting Point (clear melt)
141.0-147.0?C
Assay (GC)
≥97.5%

Description

Possesses antioxidant properties

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text