Thermo Scientific Chemicals

Furan, 99%, stab. with ca 250ppm BHT, Thermo Scientific Chemicals

Catalog number: A13102.AP
500 mL, Each
Thermo Scientific Chemicals

Furan, 99%, stab. with ca 250ppm BHT, Thermo Scientific Chemicals

Catalog number: A13102.AP
500 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A13102.AP
also known as A13102-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-00-9
IUPAC Name
furan
Molecular Formula
C4H4O
InChI Key
YLQBMQCUIZJEEH-UHFFFAOYSA-N
SMILES
O1C=CC=C1
Appearance (Color)
Clear colorless to yellow
Peroxide content
≤0.015% (as H2O2) (non-U.S. specification)
Assay (GC)
≥98.5%
Comment
≤0.2% (Methylfuran) by GC (non-U.S. specification)
Identification (FTIR)
Conforms

Description

Furan has an important biological properties and pharmacological activity. Its iodinated lipophilic derivative is widely used in the treatment of ventricular and arterial fibrillation. Its derivatives are used as inhibitors of biofilm formation for several bacterial species, as well as possess quorum-sensing inhibitory activities. 2-methylfuran and 2,5-dimethylfuran are used as lignocellulosic biofuels. Its resin is used to stop sand production in underground gas storage wells. It undergo Diels-Alder reaction with arynes to give derivatives of dihydronaphthalenes which are useful intermediates in synthesis of other polycyclic aromatic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furan has an important biological properties and pharmacological activity. Its iodinated lipophilic derivative is widely used in the treatment of ventricular and arterial fibrillation. Its derivatives are used as inhibitors of biofilm formation for several bacterial species, as well as possess quorum-sensing inhibitory activities. 2-methylfuran and 2,5-dimethylfuran are used as lignocellulosic biofuels. Its resin is used to stop sand production in underground gas storage wells. It undergo Diels-Alder reaction with arynes to give derivatives of dihydronaphthalenes which are useful intermediates in synthesis of other polycyclic aromatic compounds.

Solubility
Slightly soluble in water. Soluble in common organic solvents, including alcohol, ether and acetone.

Notes
Light, heat and air sensitive. Incompatible with oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text