Thermo Scientific Chemicals

Dimethyl carbonate, 99%, Thermo Scientific Chemicals

Catalog number: A13104.36
500 g, Each
Thermo Scientific Chemicals

Dimethyl carbonate, 99%, Thermo Scientific Chemicals

Catalog number: A13104.36
500 g, Each
Quantity
Catalog number: A13104.36
also known as A13104-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
616-38-6
IUPAC Name
dimethyl carbonate
Molecular Formula
C3H6O3
InChI Key
IEJIGPNLZYLLBP-UHFFFAOYSA-N
SMILES
COC(=O)OC
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.3670-1.3705 @ 20?C
Form
Liquid

Description

Dimethyl carbonate is used as a solvent in organic synthesis and considered as a replacement for solvent like methyl ethyl ketone, tert-butyl acetate and parachlorobenzotrifluoride. It is involved as an intermediate in the preparation of diphenylcarbonate, which in turn is used as a key raw material for the synthesis of Bisphenol-A-polycarbonate. It is also used as a 'green' methylating agent involved in the methylation of aniline, phenols and carboxylic acids. It can be used as a fuel additive due to its high oxygen content. It also finds applications related to supercapacitors and lithium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl carbonate is used as a solvent in organic synthesis and considered as a replacement for solvent like methyl ethyl ketone, tert-butyl acetate and parachlorobenzotrifluoride. It is involved as an intermediate in the preparation of diphenylcarbonate, which in turn is used as a key raw material for the synthesis of Bisphenol-A-polycarbonate. It is also used as a ′green′ methylating agent involved in the methylation of aniline, phenols and carboxylic acids. It can be used as a fuel additive due to its high oxygen content. It also finds applications related to supercapacitors and lithium batteries.

Solubility
Miscible with alcohol and ether. Immiscible with water.

Notes
Air and moisture sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, strong acids and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text