D-(+)-Glucono-1,5-lactone, 99%
D-(+)-Glucono-1,5-lactone, 99%
D-(+)-Glucono-1,5-lactone, 99%
D-(+)-Glucono-1,5-lactone, 99%
Thermo Scientific™

D-(+)-Glucono-1,5-lactone, 99%

CAS: 90-80-2 | C6H10O6 | 178.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
2500 g
Catalog number A13105.30
also known as A13105-30
Price (EUR)
22,27
Online Exclusive
26,90
Save 4,63 (17%)
Each
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (EUR)
22,27
Online Exclusive
26,90
Save 4,63 (17%)
Each
Add to cart
Chemical Identifiers
CAS90-80-2
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Assay from Suppliers CofA≥98.5% (U.S. specification)
Identification (FTIR)Conforms
Assay (Aqueous acid-base Titration)≥98.5% to ≤101.5% (non-U.S. specification)
Melting Point (clear melt)150-158°C
View more
Ask our AI about this Product
Glucono delta-lactone (GDL) is a kind of excellent multifunctional food additive. It is mainly used as protein coagulant, acidifier, pickling, or leavening agent. expander, preservative, seasonings, sequestrant, chelating agent, color-preserver, etc. it is widely applied to food, daily chemical industry, pharmaceuticals, cosmetics. Plastic and resinous modification, electrode position and polish, metal-surface cleaning and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glucono delta-lactone (GDL) is a kind of excellent multifunctional food additive. It is mainly used as protein coagulant, acidifier, pickling, or leavening agent. expander, preservative, seasonings, sequestrant, chelating agent, color-preserver, etc. it is widely applied to food, daily chemical industry, pharmaceuticals, cosmetics. Plastic and resinous modification, electrode position and polish, metal-surface cleaning and organic synthesis.

Solubility
Soluble in water 500 g/L (20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep awy from strong oxidizing agents, strong bases.
RUO – Research Use Only

General References:

  1. ET Reese.; FW Parrish.; M Ettlinger. Nojirimycin and D-glucono-1, 5-lactone as inhibitors of carbohydrases. Carbohydrate Research. 1971, 18 (3), 381-388.
  2. E DH Leaback. On the inhibition of β-N-acetyl-d-glucosaminidase by 2-acetamido-2-deoxy-d-glucono-(1→ 5)-lactone. Biochemical and biophysical research communications. 1968, 32 (6), 1025-1030.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link