Thermo Scientific Chemicals

D-(+)-Glucono-1,5-lactone, 99%, Thermo Scientific Chemicals

Catalog number: A13105.36
500 g, Each
Thermo Scientific Chemicals

D-(+)-Glucono-1,5-lactone, 99%, Thermo Scientific Chemicals

Catalog number: A13105.36
500 g, Each
Quantity
Catalog number: A13105.36
also known as A13105-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
90-80-2
Form
Powder or crystals or crystalline powder
Assay from Suppliers CofA
≥98.5% (U.S. specification)
Identification (FTIR)
Conforms
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥98.5% to ≤101.5% (non-U.S. specification)

Description

Glucono delta-lactone (GDL) is a kind of excellent multifunctional food additive. It is mainly used as protein coagulant, acidifier, pickling, or leavening agent. expander, preservative, seasonings, sequestrant, chelating agent, color-preserver, etc. it is widely applied to food, daily chemical industry, pharmaceuticals, cosmetics. Plastic and resinous modification, electrode position and polish, metal-surface cleaning and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glucono delta-lactone (GDL) is a kind of excellent multifunctional food additive. It is mainly used as protein coagulant, acidifier, pickling, or leavening agent. expander, preservative, seasonings, sequestrant, chelating agent, color-preserver, etc. it is widely applied to food, daily chemical industry, pharmaceuticals, cosmetics. Plastic and resinous modification, electrode position and polish, metal-surface cleaning and organic synthesis.

Solubility
Soluble in water 500 g/L (20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep awy from strong oxidizing agents, strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text