Sarcosine ethyl ester hydrochloride, It is used as intermediate in the synthesis of antienzyme agents for toothpaste. Found to be a marker for prostate cancer bioagression. It can be used to produce N-methyl-N-nitroso-glycine ethyl ester. This reaction will need reagent NaNO2, and the menstruum H2O and aq. acetic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sarcosine ethyl ester hydrochloride, It is used as intermediate in the synthesis of antienzyme agents for toothpaste. Found to be a marker for prostate cancer bioagression. It can be used to produce N-methyl-N-nitroso-glycine ethyl ester. This reaction will need reagent NaNO2, and the menstruum H2O and aq. acetic acid.
Solubility
Soluble in water 0.1 g/mL.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only
SI Mizushima.; T Shimanouchi.; M Tsuboi. Near Infrared Spectra of Compounds with Two Peptide Bonds and the Configuration of a Polypeptide Chain. VI. Further Evidence of the Internal Hydrogen Bonding and an Estimation of its Energy. J. Am. Chem. Soc. 1957, 79 (20), 5357-5361.
LG Marzilli.; DA Buckingham. The stereochemistry of some cobalt (III) triethylenetetramine complexes of glycine andsarcosine. Inorganic Chemistry. 1967, 6 (5), 1042-1052.