Thermo Scientific Chemicals

Chlorodimethylsilane, 97%, Thermo Scientific Chemicals

Catalog number: A13113.AK
250 mL, Each
Thermo Scientific Chemicals

Chlorodimethylsilane, 97%, Thermo Scientific Chemicals

Catalog number: A13113.AK
250 mL, Each
Quantity
Catalog number: A13113.AK
also known as A13113-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1066-35-9
IUPAC Name
chlorodimethylsilyl
Molecular Formula
C2H6ClSi
InChI Key
QABCGOSYZHCPGN-UHFFFAOYSA-N
SMILES
C[Si](C)Cl
Assay (GC)
≥96.0%
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear to slightly hazy colorless to pale yellow
Comment
Haziness may settle as precipitate on storage but will not affect performance

Description

Chlorodimethylsilane is used to prepare vinyldimethylsilanols for palladium-catalyzed cross-coupling with aryl iodides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorodimethylsilane is used to prepare vinyldimethylsilanols for palladium-catalyzed cross-coupling with aryl iodides.

Solubility
Miscible with organic solvents.

Notes
It is incompatible with strong oxidizing agents, strong acids, strong bases, metals and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text