N-(4-Bromophenyl)maleimide, 98%, Thermo Scientific Chemicals
N-(4-Bromophenyl)maleimide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-(4-Bromophenyl)maleimide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number A13119.14
also known as A13119-14
Price (USD)/ Each
277.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
277.00
Add to cart
N-(4-Bromophenyl)maleimide, 98%, Thermo Scientific Chemicals
Catalog numberA13119.14
Price (USD)/ Each
277.00
-
Add to cart
Chemical Identifiers
CAS13380-67-1
IUPAC Name1-(4-bromophenyl)-2,5-dihydro-1H-pyrrole-2,5-dione
Molecular FormulaC10H6BrNO2
InChI KeyFECSFBYOMHWJQG-UHFFFAOYSA-N
SMILESBrC1=CC=C(C=C1)N1C(=O)C=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)126.0-132.0?C
Assay (GC)≥97.5%
Appearance (Color)Pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only