3-Mercapto-1,2,4-triazole was used in a study to design a surface enhanced Raman scattering based probe for fast and accurate detection of DNA markers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Mercapto-1,2,4-triazole was used in a study to design a surface enhanced Raman scattering based probe for fast and accurate detection of DNA markers.
Solubility
Soluble in hot water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
V.Krishnakumar; R.John Xavier. FT Raman and FT-IR spectral studies of 3-mercapto-1,2,4-triazole. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2004, 60, (3),709-714
H.N.Awadalla; N.A.Hammouda; N.A.Shaaban; M.Youssef; N.N.Atta; S.T El Mansoury. A histochemical study of muscles of mice infected with Trichinella spiralis. Journal of the Egyptian Society of Parasitology. 1982, 12, (1),33-39