Thermo Scientific Chemicals

1-Chloro-3-fluorobenzene, 99%, Thermo Scientific Chemicals

Catalog number: A13123.22
100 g, Each
Thermo Scientific Chemicals

1-Chloro-3-fluorobenzene, 99%, Thermo Scientific Chemicals

Catalog number: A13123.22
100 g, Each
Quantity
Catalog number: A13123.22
also known as A13123-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
625-98-9
Identification (FTIR)
Conforms
Refractive Index
1.4925-1.4965 @ 20°C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥98.5%

Description

1-Chloro-3-fluorobenzene used as medicine and liquid crystal intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-3-fluorobenzene used as medicine and liquid crystal intermediates.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text