Thermo Scientific Chemicals

5-Chloroisatin, 98%, Thermo Scientific Chemicals

Catalog number: A13125.14
25 g, Each
Thermo Scientific Chemicals

5-Chloroisatin, 98%, Thermo Scientific Chemicals

Catalog number: A13125.14
25 g, Each
Quantity
Catalog number: A13125.14
also known as A13125-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17630-76-1
Assay (HPLC)
≥ 97.5%
Appearance (Color)
Orange to brown
Form
Powder

Description

5-Chloroisatin is a reagent used in the synthesis of Schiff bases through the reaction with 2-methyl-4-nitroaniline or 4-amino-4,5-dihydro-1H-1,2,3-triazole-5-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloroisatin is a reagent used in the synthesis of Schiff bases through the reaction with 2-methyl-4-nitroaniline or 4-amino-4,5-dihydro-1H-1,2,3-triazole-5-one.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text