Thermo Scientific Chemicals

Methyl acrylate, 99%, stab. with ca 15ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: A13128.AP
500 mL, Each
Thermo Scientific Chemicals

Methyl acrylate, 99%, stab. with ca 15ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: A13128.AP
500 mL, Each
Quantity
Catalog number: A13128.AP
also known as A13128-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
96-33-3
IUPAC Name
methyl prop-2-enoate
Molecular Formula
C4H6O2
InChI Key
BAPJBEWLBFYGME-UHFFFAOYSA-N
SMILES
COC(=O)C=C
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Assay (GC)
≥98.5%
Refractive Index
1.4010-1.4045 @ 20?C
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl acrylate is a monomer used in the preparation of polymers having rigid, flexible, ionic, nonionic, hydrophobic, or hydrophilic properties. It is involved in the production of acrylate fiber, which is used to weave synthetic carpets. It plays an important role as a reagent in the synthesis of pharmaceutical intermediates and alfa ω-dicarboxylic acid esters. It is a good nucleophile and a michael acceptor to prepare polyamidoamine (PAMAM) dendrimers.

Solubility
Miscible with water.

Notes
Light sensitive. Incompatible with oxidizing agents and peroxides.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text