Thermo Scientific Chemicals

2-Bromo-m-xylene, 98+%, Thermo Scientific Chemicals

Catalog number: A13130.30
250 g, Each
Thermo Scientific Chemicals

2-Bromo-m-xylene, 98+%, Thermo Scientific Chemicals

Catalog number: A13130.30
250 g, Each
Quantity
Catalog number: A13130.30
also known as A13130-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
576-22-7
IUPAC Name
2-bromo-1,3-dimethylbenzene
Molecular Formula
C8H9Br
InChI Key
MYMYVYZLMUEVED-UHFFFAOYSA-N
SMILES
CC1=CC=CC(C)=C1Br
Assay (GC)
≥98.0%
Refractive Index
1.5530-1.5570 @ 20?C
Appearance (Color)
Clear colorless to yellow
Identification (FTIR)
Conforms
Form
Liquid

Description

2-Bromo-m-xylene has been used to prepare 2,2,4,6,6-pentamethylbiphenyl. It is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-m-xylene has been used to prepare 2,2′,4,6,6′-pentamethylbiphenyl. It is used as an intermediate in organic synthesis.

Solubility
Soluble in aqueous solution. Very soluble in ethanol, soluble in acetone and benzene.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents. Should be stored in a tightly closed container. Store in a cool, dry, well-ventilated area.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text