Thermo Scientific Chemicals

4-Hydroxybenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: A13132.18
50 g, Each
Thermo Scientific Chemicals

4-Hydroxybenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: A13132.18
50 g, Each
Quantity
Catalog number: A13132.18
also known as A13132-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
623-05-2
IUPAC Name
4-(hydroxymethyl)phenol
Molecular Formula
C7H8O2
InChI Key
BVJSUAQZOZWCKN-UHFFFAOYSA-N
SMILES
OCC1=CC=C(O)C=C1
Appearance (Color)
White to cream to pale yellow
Assay (Silylated GC)
≥98.5%
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥98.5%

Description

4-Hydroxybenzyl alcohol is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzyl alcohol is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water (6.7 mg/ml at 20°C), dioxane (100 mg/ml), 1N NaOH (50 mg/ml), DMSO, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acid chlorides, acid anhydrides, bases, oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text