Thermo Scientific Chemicals

Dibenzo-18-crown-6, 98+%, Thermo Scientific Chemicals

Catalog number: A13133.06
5 g, Each
Thermo Scientific Chemicals

Dibenzo-18-crown-6, 98+%, Thermo Scientific Chemicals

Catalog number: A13133.06
5 g, Each
Quantity
Catalog number: A13133.06
also known as A13133-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14187-32-7
IUPAC Name
2,5,8,15,18,21-hexaoxatricyclo[20.4.0.0⁹,¹⁴]hexacosa-1(26),9,11,13,22,24-hexaene
Molecular Formula
C20H24O6
InChI Key
YSSSPARMOAYJTE-UHFFFAOYSA-N
SMILES
C1COC2=CC=CC=C2OCCOCCOC2=CC=CC=C2OCCO1
Form
Powder
Appearance (Color)
White to pale cream to cream to green
Identification (FTIR)
Conforms
Assay (HPLC)
≥98.0%
Assay (GC)
≥98.0%

Description

Dibenzo-18-crown-6, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is an important organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzo-18-crown-6, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is an important organic intermediate.

Solubility
Sparingly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible to strong oxidizing agents, strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text