Thermo Scientific Chemicals

2'-Hydroxy-5'-methylacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: A13136.14
25 g, Each
Thermo Scientific Chemicals

2'-Hydroxy-5'-methylacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: A13136.14
25 g, Each
Quantity
Catalog number: A13136.14
also known as A13136-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1450-72-2
IUPAC Name
1-(2-hydroxy-5-methylphenyl)ethan-1-one
Molecular Formula
C9H10O2
InChI Key
YNPDFBFVMJNGKZ-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC(C)=CC=C1O
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Melting Point (clear melt)
44.0-50.0?C
Appearance (Color)
Pale yellow

Description

2'-Hydroxy-5'-methylacetophenone is used to produce 2-ethyl-4-methyl-phenol. It is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Hydroxy-5′-methylacetophenone is used to produce 2-ethyl-4-methyl-phenol. It is used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text