Thermo Scientific Chemicals

N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%, Thermo Scientific Chemicals

Catalog number: A13141.06
5 g, Each
Thermo Scientific Chemicals

N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%, Thermo Scientific Chemicals

Catalog number: A13141.06
5 g, Each
Quantity
Catalog number: A13141.06
also known as A13141-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
24589-78-4
IUPAC Name
2,2,2-trifluoro-N-methyl-N-(trimethylsilyl)acetamide
Molecular Formula
C6H12F3NOSi
InChI Key
MSPCIZMDDUQPGJ-UHFFFAOYSA-N
SMILES
CN(C(=O)C(F)(F)F)[Si](C)(C)C
Form
Liquid
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Refractive Index
1.3775-1.3825 @ 20?C

Description

N-Methyl-N-(trimethylsilyl)trifluoroacetamide is used as a silylating reagent that forms volatile derivatives for GC-MS analysis. It acts as a pharmaceutical intermediate and metabolite of Famprofazone in humans. It is also used in the detection of steroid hormones 17-alfa-ethynylestradiol and estrone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methyl-N-(trimethylsilyl)trifluoroacetamide is used as a silylating reagent that forms volatile derivatives for GC-MS analysis. It acts as a pharmaceutical intermediate and metabolite of Famprofazone in humans. It is also used in the detection of steroid hormones 17-alfa-ethynylestradiol and estrone.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text