Thermo Scientific Chemicals

1,2-Phenylenediacetonitrile, 98%, Thermo Scientific Chemicals

Catalog number: A13142.06
5 g, Each
Thermo Scientific Chemicals

1,2-Phenylenediacetonitrile, 98%, Thermo Scientific Chemicals

Catalog number: A13142.06
5 g, Each
Quantity
Catalog number: A13142.06
also known as A13142-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
613-73-0
IUPAC Name
2-[2-(cyanomethyl)phenyl]acetonitrile
Molecular Formula
C10H8N2
InChI Key
FWPFXBANOKKNBR-UHFFFAOYSA-N
SMILES
N#CCC1=CC=CC=C1CC#N
Assay (GC)
≥97.5%
Melting Point (clear melt)
55.5-61.5?C
Appearance (Color)
White to pale yellow to pale brown
Form
Crystals or powder or crystalline powder

Description

1,2-Phenylenediacetonitrile is used as API and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Phenylenediacetonitrile is used as API and intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text