2-Mercapto-4-methylpyrimidine hydrochloride, 99%, Thermo Scientific Chemicals
2-Mercapto-4-methylpyrimidine hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Mercapto-4-methylpyrimidine hydrochloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A13145.14
also known as A13145-14
Price (USD)/ Each
35.50
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
35.50
Add to cart
2-Mercapto-4-methylpyrimidine hydrochloride, 99%, Thermo Scientific Chemicals
Catalog numberA13145.14
Price (USD)/ Each
35.50
-
Add to cart
Chemical Identifiers
CAS6959-66-6
IUPAC Name6-methyl-1,2-dihydropyrimidine-2-thione hydrochloridyl
Molecular FormulaC5H6ClN2S
InChI KeyBQUGSWHPLSUHNI-UHFFFAOYSA-N
SMILES[Cl].CC1=CC=NC(=S)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormPowder
Assay (Titration ex Chloride)≥98.5 to ≤101.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only