Thermo Scientific Chemicals

4-Bromoisophthalic acid, 96%, Thermo Scientific Chemicals

Catalog number: A13152.14
25 g, Each
Thermo Scientific Chemicals

4-Bromoisophthalic acid, 96%, Thermo Scientific Chemicals

Catalog number: A13152.14
25 g, Each
Quantity
Catalog number: A13152.14
also known as A13152-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6939-93-1
IUPAC Name
4-bromobenzene-1,3-dicarboxylic acid
Molecular Formula
C8H5BrO4
InChI Key
MSQIEZXCNYUWHN-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(Br)C(=C1)C(O)=O
Form
Powder or lumps
Assay (Aqueous acid-base Titration)
≥95.0 to ≤105.0%
Assay (Silylated GC)
≥95.0%
Melting Point (clear melt)
292-305°C
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text