DL-Ethionine, 98%, Thermo Scientific Chemicals
DL-Ethionine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DL-Ethionine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number A13154.14
also known as A13154-14
Price (USD)/ Each
714.65
Online exclusive
794.00 
Save 79.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
714.65
Online exclusive
794.00 
Save 79.35 (10%)
Add to cart
DL-Ethionine, 98%, Thermo Scientific Chemicals
Catalog numberA13154.14
Price (USD)/ Each
714.65
Online exclusive
794.00 
Save 79.35 (10%)
-
Add to cart
Chemical Identifiers
CAS67-21-0
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps or flakes
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Appearance (Color)White to cream
DL-Ethionine is used as a dietary supplement to accelerate cholangiocarcinogenesis in vivo. It is also used to induce oxidative stress in liver to study the levels and activities of anti-oxidative enzymes and compounds such as glutathione.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Liu, Q.; Liang, Y.; Zhang, Y.; Shang, X.; Liu, S.; Wen, J.; Wen, T. YjeH Is a Novel Exporter of l-Methionine and Branched-Chain Amino Acids in Escherichia coli. Appl. Environ. Microbiol. 2015, 81 (22), 7753-7766.
  2. Ceranowicz, P.; Cieszkowski, J.; Warzecha, Z.; Dembiński, A. Eksperymentalne modele ostrego zapalenia trzustki. Postepy. Hig. Med. Dosw. 2015, 69, 264-269.