Thermo Scientific Chemicals

1,10-Phenanthroline, 99%, may contain up to 1.5% water, Thermo Scientific Chemicals

Catalog number: A13163.14
25 g, Each
Thermo Scientific Chemicals

1,10-Phenanthroline, 99%, may contain up to 1.5% water, Thermo Scientific Chemicals

Catalog number: A13163.14
25 g, Each
Quantity
Catalog number: A13163.14
also known as A13163-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
66-71-7
IUPAC Name
1,10-phenanthroline
Molecular Formula
C12H8N2
InChI Key
DGEZNRSVGBDHLK-UHFFFAOYSA-N
SMILES
C1=CN=C2C(C=CC3=CC=CN=C23)=C1
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5% (dry wt. basis)
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤1.5%
Melting Point (clear melt)
113.0-121.0?C
Appearance (Color)
White to cream

Description

1,10-Phenanthroline, anhydrous is used as a chelating agent, used as bidendate nitrogen ligands in Heck type reactions as an alternative to phospine ligands. It is widely used as an indicator for alkyllithium reagents and an inhibitor of metallopeptidase enzymes. It is also used in the analysis formulations in end-user industries like food, beverage and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Phenanthroline is used as a chelating agent, used as bidendate nitrogen ligands in Heck type reactions as an alternative to phospine ligands. It is widely used as an indicator for alkyllithium reagents and an inhibitor of metallopeptidase enzymes. It is also used in the analysis formulations in end-user industries like food, beverage and pharmaceuticals.

Solubility
Soluble in acetone and ethanol. Slightly soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids. Store in a tightly closed container. Store in a cool, dry, well-ventilated area.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text