Thermo Scientific Chemicals

2-Chloro-6-hydroxypyridine, 98%, Thermo Scientific Chemicals

Catalog number: A13164.14
25 g, Each
Thermo Scientific Chemicals

2-Chloro-6-hydroxypyridine, 98%, Thermo Scientific Chemicals

Catalog number: A13164.14
25 g, Each
Quantity
Catalog number: A13164.14
also known as A13164-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16879-02-0
IUPAC Name
6-chloro-1,2-dihydropyridin-2-one
Molecular Formula
C5H4ClNO
InChI Key
CLNNBQDAAGDAHI-UHFFFAOYSA-N
SMILES
ClC1=CC=CC(=O)N1
Appearance (Color)
White to pale yellow
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥97.5%
Melting Point (clear melt)
126.5-132.5?C

Description

6-chloro-2-hydroxypyridine was used to study the synthesis, electrochemistry, spectroscopic properties and X-ray crystal structure of Os2Cl(chp)4 (chp = 6-chloro-2- hydroxypyridinate) complex. It undergoes melt reaction with Os2(O2CCH3)4CI2 at 145°C to yield Os2CI4(chp) and Os2Cl(chp)4.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-chloro-2-hydroxypyridine was used to study the synthesis, electrochemistry, spectroscopic properties and X-ray crystal structure of Os2Cl(chp)4 (chp = 6-chloro-2- hydroxypyridinate) complex. It undergoes melt reaction with Os2(O2CCH3)4CI2 at 145°C to yield Os2CI4(chp) and Os2Cl(chp)4.

Notes
Store at room temperature. Ensure adequate ventilation. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text