6-chloro-2-hydroxypyridine was used to study the synthesis, electrochemistry, spectroscopic properties and X-ray crystal structure of Os2Cl(chp)4 (chp = 6-chloro-2- hydroxypyridinate) complex. It undergoes melt reaction with Os2(O2CCH3)4CI2 at 145°C to yield Os2CI4(chp) and Os2Cl(chp)4.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-chloro-2-hydroxypyridine was used to study the synthesis, electrochemistry, spectroscopic properties and X-ray crystal structure of Os2Cl(chp)4 (chp = 6-chloro-2- hydroxypyridinate) complex. It undergoes melt reaction with Os2(O2CCH3)4CI2 at 145°C to yield Os2CI4(chp) and Os2Cl(chp)4.
Notes
Store at room temperature. Ensure adequate ventilation. Incompatible with oxidizing agents.
RUO – Research Use Only
Daniel L. Comins.; Hao Hong.; Gao Jianhua. Asymmetric synthesis of camptothecin alkaloids: A nine-step synthesis of (S)-camptothecin. Tetrahedron Lett. 1994, 35 (30), 5331-5334 .
Daniel L. Comins.; Jason M. Nolan. A Practical Six-Step Synthesis of (S)-Camptothecin.Org. Lett. 2001, 3 (26),4255-4257 .