Thermo Scientific Chemicals

Adipoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A13168.36
500 g, Each
Thermo Scientific Chemicals

Adipoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A13168.36
500 g, Each
Quantity
Catalog number: A13168.36
also known as A13168-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-50-2
IUPAC Name
hexanedioyl dichloride
Molecular Formula
C6H8Cl2O2
InChI Key
PWAXUOGZOSVGBO-UHFFFAOYSA-N
SMILES
ClC(=O)CCCCC(Cl)=O
Appearance (Color)
Clear colorless to yellow to brown to dark brown
Assay (GC)
≥97.5%
Refractive Index
1.4670-1.4730 @ 20?C
Form
Liquid
Comment
May darken on storage

Description

Adipoyl chloride is used in the production of nylon by reacting with hexamethylene diamine. It is also used in the preparation of biphenyl end-capped liquid crystals. Further, it is employed in the synthesis of chiral polymer for membrane applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adipoyl chloride is used in the production of nylon by reacting with hexamethylene diamine. It is also used in the preparation of biphenyl end-capped liquid crystals. Further, it is employed in the synthesis of chiral polymer for membrane applications.

Solubility
Immiscible with water.

Notes
Incompatible strong oxidizing agents, bases and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text